Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163486.1 | NP_001156958.1 | 5, 6 | 776 | ||
NM_198030.2 | NP_932147.2 | 5, 6 | 776 | |||
GenBank mRNA | AY101768.1 | - | 5, 6 | 703 | ||
BC019427.1 | - | 5, 6 | 752 | 5132342 | ||
DV660487.1 | - | 2, 3 | 202 |