Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159421.1 | NP_001152893.1 | 2 | 609 | ||
NM_001159422.1 | NP_001152894.1 | 3 | 386 | |||
NM_198645.2 | NP_941047.1 | 3 | 735 | |||
GenBank mRNA | AK135450.1 | - | 3 | 315 | ||
AV583692.1 | - | 3 | 386 | |||
BB686073.1 | - | 1 | 50 | |||
BC024406.1 | - | 3 | 299 | |||
BC024862.1 | - | 3 | 710 | 5356370 | ||
BC053690.1 | - | 3 | 287 | |||
CF617453.1 | - | 3 | 713 | |||
CJ094333.1 | - | 2 | 190 |