Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199467.2 | NP_955761.2 | 11 | 1722 | ||
GenBank mRNA | AK089531.1 | - | 4 | 462 | ||
AK166072.1 | - | 11 | 1733 | |||
AY512923.1 | - | 6 | 856 | |||
BC051674.1 | - | 2 | 169 | |||
BC094429.1 | - | 10 | 1066 | |||
BC137910.1 | - | 6 | 632 | 8860929 | ||
BC145028.1 | - | 6 | 632 | 9053576 |