Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008551.1 | NP_032577.1 | 10 | 2026 | ||
GenBank mRNA | AK171912.1 | - | 1 | 575 | ||
BC024559.1 | - | 6 | 1395 | |||
BC063064.1 | - | 10 | 2026 | 5687007 |