Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010078.3 | NP_034208.2 | 6 | 820 | ||
GenBank mRNA | AK034383.1 | - | 6 | 763 | ||
AK081426.1 | - | 6 | 764 | |||
AK158102.1 | - | 5 | 622 | |||
BC060605.1 | - | 5 | 591 | |||
BC125345.1 | - | 6 | 680 | 40129860 | ||
BC125347.1 | - | 6 | 680 | 40129862 |