Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oc90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010953.2 | NP_035083.1 | 15 | 1298 | ||
GenBank mRNA | AF091846.1 | - | 12 | 1107 | ||
AF093591.1 | - | 15 | 1408 | |||
AK012981.1 | - | 15 | 1300 | |||
BC048716.1 | - | 13 | 1169 | 6334417 |