Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011516.2 | NP_035646.2 | 6 | 428 | ||
GenBank mRNA | AK015000.1 | - | 6 | 429 | ||
AK133229.1 | - | 6 | 429 | |||
BC137967.1 | - | 6 | 390 | 8860986 | ||
D88539.1 | - | 1 | 18 | |||
L41069.1 | - | 6 | 409 | |||
Z38118.1 | - | 6 | 419 |