Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011516.2 | NP_035646.2 | 30, 31 | 2750 | ||
GenBank mRNA | AK076780.1 | - | 9, 10 | 841 | ||
BC137967.1 | - | 30, 31 | 2712 | 8860986 | ||
L41069.1 | - | 30, 31 | 2731 | |||
Z38118.1 | - | 30, 31 | 2741 |