Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204255.1 | NP_001191184.1 | 6 | 1080 | ||
NM_005506.3 | NP_005497.1 | 9 | 1509 | |||
GenBank mRNA | AK296519.1 | - | 6 | 784 | ||
AK313016.1 | - | 9 | 1219 | |||
BC021892.1 | - | 9 | 1175 | 3872778 | ||
BT006939.1 | - | 9 | 1153 | |||
D12676.1 | - | 9 | 1404 |