Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290502.1 | NP_001277431.1 | 10 | 1708 | ||
NM_025978.4 | NP_080254.2 | 10 | 1470 | |||
GenBank mRNA | AK015308.1 | - | 10 | 1430 | ||
AK016619.1 | - | 9 | 1234 | |||
AK031605.1 | - | 10 | 1701 | |||
AK035570.1 | - | 10 | 1440 | |||
AK041095.1 | - | 10 | 1444 | |||
AK044266.1 | - | 10 | 1446 | |||
AK134539.1 | - | 10 | 1461 | |||
BC024847.1 | - | 10 | 1435 | 5355806 | ||
BC138212.1 | - | 10 | 1432 | 8861231 | ||
BC145944.1 | - | 10 | 1432 | 40131247 |