Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290476.1 | NP_001277405.1 | 3, 4 | 551 | ||
NM_026167.4 | NP_080443.1 | 5, 6 | 570 | |||
GenBank mRNA | AK004677.1 | - | 5, 6 | 542 | ||
AK122491.1 | - | 3, 4 | 551 | |||
AK191256.1 | - | 2, 3 | 277 |