Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027554.2 | NP_081830.2 | 10 | 4523 | ||
GenBank mRNA | AK129464.1 | - | 10 | 4478 | ||
AK142200.1 | - | 10 | 4519 | |||
BC054404.1 | - | 10 | 4523 | 6494448 | ||
BC057122.1 | - | 10 | 4513 | 6834543 | ||
BC058784.1 | - | 10 | 4450 | 6310341 | ||
BC066786.1 | - | 10 | 4464 |