Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi27l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029803.3 | NP_084079.1 | 4 | 353 | ||
GenBank mRNA | AK010014.1 | - | 334 | |||
AK160503.1 | - | 4 | 334 | |||
BC086889.1 | - | 4 | 333 | |||
BC100588.1 | - | 4 | 353 | 6759652 |