Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr1384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146472.2 | NP_666683.1 | 1 | 289 | ||
GenBank mRNA | BC119159.1 | - | 1 | 289 | 8733908 | |
BC120667.1 | - | 1 | 289 | 40129590 |