Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147082.2 | NP_667293.2 | 1 | 765 | ||
GenBank mRNA | BC128008.1 | - | 1 | 769 | 40135990 |