Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177765.3 | NP_808433.2 | 10, 11 | 1659 | ||
GenBank mRNA | AK029565.1 | - | 10, 11 | 1658 | ||
AK161519.1 | - | 10, 11 | 1482 | |||
AM690757.1 | - | 9, 10 | 1360 | |||
BC145749.1 | - | 9, 10 | 1267 |