Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mex3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198615.2 | NP_941017.2 | 2 | 1309 | ||
GenBank mRNA | AK149801.1 | - | 2 | 837 | ||
BC059858.1 | - | 3 | 429 |