Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Uggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198899.2 | NP_942602.2 | 28, 29 | 3287 | ||
GenBank mRNA | AK144381.1 | - | 27, 28 | 3116 | ||
BC023910.1 | - | 1, 2 | 83 | |||
BC051172.1 | - | 2, 3 | 391 | |||
BC062936.1 | - | 28, 29 | 3234 | 6838545 | ||
CD808359.1 | - | 1, 2 | 50 |