Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201226.1 | NP_957678.1 | 6, 7 | 1511 | ||
GenBank mRNA | AK013512.1 | - | 5, 6 | 488 | ||
AK129306.1 | - | 6, 7 | 1320 | |||
AK154897.1 | - | 6, 7 | 1541 | |||
AK192094.1 | - | 2, 3 | 131 | |||
BC094573.1 | - | 6, 7 | 1501 | 30354494 | ||
BC138462.1 | - | 6, 7 | 1583 | 8861482 | ||
BC138463.1 | - | 6, 7 | 1583 | 8861483 |