Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289458.1 | NP_001276387.1 | 10 | 1260 | ||
NM_001289459.1 | NP_001276388.1 | 10 | 1249 | |||
NM_001289461.1 | NP_001276390.1 | 9 | 1067 | |||
NM_010427.5 | NP_034557.3 | 10 | 1232 | |||
GenBank mRNA | AK042121.1 | - | 10 | 1178 | ||
BC119228.1 | - | 10 | 1119 | 8733977 | ||
BC144764.1 | - | 10 | 1131 | |||
D10212.1 | - | 9 | 1055 | |||
D10213.1 | - | 9 | 1040 | |||
S71816.1 | - | 9 | 1055 | |||
X72307.1 | - | 9 | 1116 | |||
X84046.1 | - | 9 | 1095 |