Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252207.1 | NP_001239136.1 | 3 | 1617 | ||
NM_001252208.1 | NP_001239137.1 | 2 | 1594 | |||
NM_001252209.2 | NP_001239138.1 | 2 | 1584 | |||
NM_001252210.2 | NP_001239139.1 | 2 | 1565 | |||
NM_010604.3 | NP_034734.3 | 2 | 1536 | |||
GenBank mRNA | AK032192.1 | - | 2 | 1550 | ||
AK045447.1 | - | 2 | 1540 | |||
AK078905.1 | - | 2 | 1537 | |||
AK128942.1 | - | 2 | 1536 | |||
AK140249.1 | - | 2 | 1547 | |||
BC033442.1 | - | 3 | 1592 | 4223640 | ||
BC092296.1 | - | 1 | 1427 | 5064724 |