Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039089.1 | NP_001034178.1 | 3 | 421 | ||
NM_011344.2 | NP_035474.1 | 3 | 421 | |||
GenBank mRNA | AF063095.1 | - | 3 | 370 | ||
AK005023.2 | - | 3 | 394 | |||
AK005544.1 | - | 3 | 404 | |||
AK146022.1 | - | 3 | 395 | |||
AK160420.1 | - | 3 | 403 | |||
AK160936.1 | - | 3 | 379 | |||
AK161546.1 | - | 3 | 395 | |||
AK220502.1 | - | 3 | 421 | |||
BC026816.1 | - | 3 | 393 | 4457637 |