Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011506.3 | NP_035636.1 | 7 | 940 | ||
GenBank mRNA | AF058955.1 | - | 6 | 807 | ||
AK005273.1 | - | 7 | 923 | |||
AK081965.1 | - | 7 | 961 | |||
AK088801.1 | - | 7 | 941 | |||
AK132762.1 | - | 7 | 930 | |||
AK150594.1 | - | 7 | 940 | |||
AK152783.1 | - | 7 | 940 | |||
AK153198.1 | - | 7 | 940 | |||
AK160130.1 | - | 7 | 941 | |||
AK160652.1 | - | 7 | 929 | |||
BC038497.1 | - | 1 | 108 | |||
BC052774.1 | - | 4 | 472 | |||
BC056353.1 | - | 7 | 931 | 6808360 | ||
BC057605.1 | - | 7 | 931 | 6808360 |