Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023750.2 | NP_076239.1 | 5 | 748 | ||
GenBank mRNA | AK014610.1 | - | 5 | 704 | ||
AK034259.1 | - | 5 | 643 | |||
AK083748.1 | - | 5 | 688 | |||
AK145426.1 | - | 5 | 749 | |||
AK158422.1 | - | 5 | 698 | |||
BC008540.1 | - | 5 | 696 | 3495627 |