Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080974.2 | NP_001074443.1 | 2, 3 | 193 | ||
NM_025618.3 | NP_079894.2 | 2, 3 | 233 | |||
GenBank mRNA | AK008404.1 | - | 2, 3 | 185 | ||
AK008783.1 | - | 2, 3 | 234 | |||
AK008970.1 | - | 2, 3 | 182 | |||
AK134747.1 | - | 2, 3 | 195 | |||
AK146071.1 | - | 2, 3 | 190 | |||
AK146911.1 | - | 2, 3 | 174 | |||
AK168572.1 | - | 2, 3 | 174 | |||
AK168792.1 | - | 2, 3 | 174 | |||
AK168900.1 | - | 2, 3 | 165 | |||
BC065790.1 | - | 2, 3 | 160 | 948616 | ||
BY708689.1 | - | 2, 3 | 231 |