Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PQBP1 SIRNA Assays ›
Gene Symbol:
PQBP1
Gene Name:
polyglutamine binding protein 1
Gene Aliases:
MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS
Chromosome Location:
Chr.X: 48897862 - 48903145 on Build GRCh38
UniGene ID:
Hs.534384
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
16600
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PQBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032381.1
NP_001027553.1
3
204
NM_001032382.1
NP_001027554.1
3
272
NM_001032383.1
NP_001027555.1
3
283
NM_001032384.1
NP_001027556.1
3
192
NM_001167989.1
NP_001161461.1
3
204
NM_001167990.1
NP_001161462.1
3
186
NM_001167992.1
NP_001161464.1
2
101
NM_005710.2
NP_005701.1
2
337
NM_144495.2
NP_652766.1
3
272
XM_005272571.3
XP_005272628.1
3
267
XM_005272572.4
XP_005272629.1
3
195
XM_011543884.2
XP_011542186.1
3
342
XM_017029207.1
XP_016884696.1
3
333
GenBank mRNA
AB016533.1
-
3
253
AB041833.1
-
2
83
AB041834.1
-
2
83
AB041835.1
-
2
83
AB041836.1
-
2
83
AJ005893.1
-
2
340
AJ242829.1
-
3
143
AJ973593.1
-
2
98
AJ973594.1
-
2
98
AJ973595.1
-
2
98
AJ973596.1
-
98
AJ973597.1
-
2
98
AJ973598.1
-
2
98
AJ973599.1
-
2
98
AJ973600.1
-
2
98
AJ973601.1
-
2
98
AJ973602.1
-
2
98
AJ973603.1
-
2
98
AJ973604.1
-
2
102
AJ973605.1
-
2
98
AJ973606.1
-
2
98
AJ973607.1
-
2
98
BC012358.1
-
3
164
4662649
BE396796.1
-
3
184
BI818141.1
-
3
166
BX362311.2
-
3
241
BX369697.2
-
3
424
CD051018.1
-
3
207
CN292761.1
-
3
272
CN292769.1
-
3
190
KU178500.1
-
2
83
KU178501.1
-
2
83
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
neuron projection development
regulation of RNA splicing
regulation of dendrite morphogenesis
cilium morphogenesis
DNA binding
transcription coactivator activity
protein binding
protein C-terminus binding
ribonucleoprotein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0