Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025963.3 | NP_080239.1 | 2 | 173 | ||
GenBank mRNA | AK002558.1 | - | 2 | 171 | ||
AK008797.1 | - | 2 | 174 | |||
AK011065.1 | - | 2 | 166 | |||
AK020219.1 | - | 2 | 174 | |||
AK136533.1 | - | 2 | 172 | |||
AK145441.1 | - | 2 | 166 | |||
AK150803.1 | - | 2 | 172 | |||
AK151703.1 | - | 2 | 172 | |||
AK157104.1 | - | 2 | 172 | |||
BC003853.1 | - | 2 | 160 | 3601013 | ||
BC019725.1 | - | 2 | 160 | 3497741 | ||
BC086919.1 | - | 2 | 172 | 6763104 | ||
BC089323.1 | - | 1 | 149 | 6814929 |