Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026157.2 | NP_080433.1 | 2 | 263 | ||
GenBank mRNA | AK002680.1 | - | 2 | 253 | ||
AK011546.1 | - | 2 | 264 | |||
AK036760.1 | - | 2 | 264 | |||
AK045203.1 | - | 2 | 253 | |||
AK076565.1 | - | 2 | 253 | |||
AK135537.1 | - | 2 | 231 | |||
AK154747.1 | - | 2 | 260 | |||
AK166038.1 | - | 2 | 260 | |||
BC017535.1 | - | 2 | 252 | |||
BC057643.1 | - | 2 | 237 | 2616306 |