Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026298.5 | NP_080574.5 | 5, 6 | 512 | ||
GenBank mRNA | AK043642.1 | - | 5, 6 | 460 | ||
AK144190.1 | - | 5, 6 | 439 | |||
AY339616.1 | - | 5, 6 | 448 | |||
BC042583.1 | - | 5, 6 | 452 | |||
BC066096.1 | - | 5, 6 | 505 | 6856438 | ||
BU706379.1 | - | 5, 6 | 512 |