Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114977.1 | NP_001108449.1 | 5 | 480 | ||
NM_026476.2 | NP_080752.2 | 4 | 348 | |||
GenBank mRNA | AK011588.1 | - | 5 | 368 | ||
AK011796.1 | - | 5 | 370 | |||
AK029898.1 | - | 5 | 379 | |||
BC031497.1 | - | 5 | 343 | |||
BC068265.1 | - | 4 | 364 | 30022109 | ||
BC080699.1 | - | 4 | 231 |