Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005717.3 | NP_005708.1 | 1, 2 | 435 | ||
GenBank mRNA | AF006088.1 | - | 1, 2 | 177 | ||
AF017807.1 | - | 1, 2 | 156 | |||
AK129598.1 | - | 1, 2 | 220 | |||
BI758106.1 | - | 1, 2 | 344 | |||
CN291383.1 | - | 1, 2 | 435 |