Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109626.1 | NP_001103096.1 | 1 | 200 | ||
NM_001109628.1 | NP_001103098.1 | 1 | 200 | |||
NM_026952.2 | NP_081228.2 | 1 | 200 | |||
GenBank mRNA | AA066711.1 | - | 1 | 201 | ||
AK007584.1 | - | 1 | 63 | |||
AK129237.1 | - | 1 | 153 | |||
AK164881.1 | - | 1 | 64 | |||
AY072295.1 | - | 1 | 191 | |||
BC050142.1 | - | 1 | 170 | |||
BC116645.1 | - | 1 | 183 | 40086381 |