Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027135.2 | NP_081411.2 | 6 | 785 | ||
GenBank mRNA | BC057981.1 | - | 4 | 479 | ||
BC067020.1 | - | 6 | 785 | 6847085 |