Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277140.1 | NP_001264069.1 | 3 | 259 | ||
NM_005721.4 | NP_005712.1 | 3 | 468 | |||
GenBank mRNA | AF006083.1 | - | 3 | 294 | ||
AF127773.1 | - | 3 | 337 | |||
AK302152.1 | - | 3 | 213 | |||
AK302359.1 | - | 3 | 247 | |||
AK303988.1 | - | 3 | 305 | |||
AK312659.1 | - | 3 | 268 | |||
AK316213.1 | - | 3 | 290 | |||
AK316396.1 | - | 3 | 259 | |||
BC044590.1 | - | 3 | 442 | 5261640 | ||
DB453995.1 | - | 3 | 468 |