Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029714.3 | NP_083990.2 | 19 | 2295 | ||
GenBank mRNA | AK077059.1 | - | 19 | 2277 | ||
AK138347.1 | - | 14 | 1892 | |||
BC144969.1 | - | 19 | 2275 | |||
GQ225581.1 | - | 18 | 2196 |
Gene Symbol: Catsperg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164658.1 | NP_001158130.1 | 18 | 2166 | ||
GenBank mRNA | AK039202.1 | - | 11 | 1592 | ||
AK133307.1 | - | 18 | 1947 | |||
BC172039.1 | - | 12 | 1206 |