Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282704.1 | NP_001269633.1 | 6 | 773 | ||
NM_001282705.1 | NP_001269634.1 | 5 | 795 | |||
NM_001282706.1 | NP_001269635.1 | 5 | 625 | |||
NM_001282707.1 | NP_001269636.1 | 5 | 600 | |||
NM_005742.3 | NP_005733.1 | 4 | 422 | |||
XM_011510308.1 | XP_011508610.1 | 4 | 363 | |||
GenBank mRNA | AK127433.1 | - | 6 | 773 | ||
AK131234.1 | - | 5 | 791 | |||
AK225496.1 | - | 3 | 168 | |||
AK289428.1 | - | 4 | 298 | |||
AK294347.1 | - | 5 | 515 | |||
AK297547.1 | - | 5 | 585 | |||
BC001312.1 | - | 4 | 378 | 3454007 | ||
BF979486.1 | - | 427 | ||||
D49489.1 | - | 373 | ||||
HY166449.1 | - | 4 | 413 | |||
U79278.1 | - | 3 | 224 |