Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153805.1 | NP_722500.1 | 6 | 758 | ||
GenBank mRNA | AK142182.1 | - | 5 | 1274 | ||
AK142988.1 | - | 5 | 1330 | |||
AK154801.1 | - | 5 | 1296 | |||
BC034126.1 | - | 6 | 758 | 4912536 |