Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175472.3 | NP_780681.2 | 13 | 2498 | ||
GenBank mRNA | AK031299.1 | - | 11 | 1891 | ||
AK049279.1 | - | 11 | 1487 | |||
AK220327.1 | - | 11 | 1746 | |||
BC145507.1 | - | 11 | 2079 | |||
BC150791.1 | - | 12 | 2149 | 9087702 |