Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146010.1 | NP_001139482.1 | 8 | 835 | ||
NM_199012.2 | NP_950177.2 | 8 | 835 | |||
GenBank mRNA | AK122368.1 | - | 8 | 727 | ||
AK140322.1 | - | 8 | 814 | |||
AK141949.1 | - | 8 | 835 | |||
AK156841.1 | - | 7 | 661 | |||
AK157054.1 | - | 8 | 808 | |||
BC016532.1 | - | 8 | 834 | |||
BC060647.1 | - | 8 | 797 | 6854041 |