Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201607.2 | NP_963901.1 | 3, 4 | 421 | ||
GenBank mRNA | AK149514.1 | - | 3, 4 | 371 | ||
BC030873.1 | - | 3, 4 | 421 | 4223352 |