Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206973.2 | NP_996856.1 | 2 | 1674 | ||
GenBank mRNA | AK044225.1 | - | 2 | 1731 | ||
AK044366.1 | - | 2 | 1689 | |||
BC099681.1 | - | 2 | 1674 | 6494327 |
Set Membership: |
![]() |