Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Exoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207214.3 | NP_997097.1 | 11, 12 | 1667 | ||
GenBank mRNA | AK049416.1 | - | 1, 2 | 592 | ||
AK134562.1 | - | 11, 12 | 1180 | |||
AK152307.1 | - | 11, 12 | 1396 | |||
AK157473.1 | - | 11, 12 | 1362 | |||
AK164067.1 | - | 11, 12 | 1389 | |||
AK167145.1 | - | 11, 12 | 1355 | |||
AK170148.1 | - | 11, 12 | 1361 | |||
AK171012.1 | - | 11, 12 | 1385 | |||
BC049967.1 | - | 11, 12 | 1324 | 3672313 |