Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166493.1 | NP_001159965.1 | 17 | 3288 | ||
NM_207246.4 | NP_997129.1 | 18 | 3280 | |||
GenBank mRNA | AK173041.1 | - | 17 | 3173 | ||
BC066069.1 | - | 16 | 2918 | 6851971 | ||
BC066127.1 | - | 18 | 3205 | 30280332 |