Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC052040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145898.1 | NP_001139370.1 | 6, 7 | 594 | ||
NM_207264.4 | NP_997147.3 | 6, 7 | 594 | |||
GenBank mRNA | AK090080.1 | - | 6, 7 | 534 | ||
AK154260.1 | - | 6, 7 | 572 | |||
AK167625.1 | - | 6, 7 | 552 | |||
BC052040.1 | - | 6, 7 | 577 | 5718089 |