Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099282.1 | NP_001092752.1 | 4 | 603 | ||
NM_001099283.1 | NP_001092753.1 | 2 | 440 | |||
NM_001099284.1 | NP_001092754.1 | 3 | 563 | |||
NM_001324347.1 | NP_001311276.1 | 3 | 481 | |||
NM_001324348.1 | NP_001311277.1 | 2 | 509 | |||
NM_001324349.1 | NP_001311278.1 | 4 | 673 | |||
NM_001324350.1 | NP_001311279.1 | 3 | 550 | |||
NM_001324351.1 | NP_001311280.1 | 3 | 632 | |||
NM_001324352.1 | NP_001311281.1 | 4 | 608 | |||
NM_001324353.1 | NP_001311282.1 | 5 | 713 | |||
NM_005674.2 | NP_005665.2 | 2 | 922 | |||
XM_005271832.2 | XP_005271889.1 | 2 | 435 | |||
XM_006718001.2 | XP_006718064.1 | 2 | 534 | |||
XM_006718003.3 | XP_006718066.1 | 2 | 434 | |||
XM_011540232.2 | XP_011538534.1 | 3 | 3521 | |||
XM_011540234.2 | XP_011538536.1 | 3 | 568 | |||
XM_011540235.2 | XP_011538537.1 | 3 | 567 | |||
XM_011540236.2 | XP_011538538.1 | 3 | 576 | |||
XM_011540237.2 | XP_011538539.1 | 3 | 526 | |||
XM_011540238.2 | XP_011538540.1 | 2 | 378 | |||
XM_017016740.1 | XP_016872229.1 | 4 | 605 | |||
GenBank mRNA | AK292013.1 | - | 3 | 559 | ||
BC026030.1 | - | 4 | 547 | 5460088 | ||
DA686973.1 | - | 2 | 436 | |||
DA937379.1 | - | 3 | 559 | |||
DN994387.1 | - | 3 | 608 | |||
X82125.1 | - | 2 | 922 |