Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010069.1 | NP_034199.1 | 2 | 916 | ||
GenBank mRNA | BC055768.1 | - | 2 | 916 | 6413208 | |
D50000.1 | - | 1 | 339 |