Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010143.1 | NP_034273.1 | 3 | 570 | ||
GenBank mRNA | AK144414.1 | - | 3 | 586 | ||
AK158043.1 | - | 3 | 1631 | |||
BC014822.1 | - | 3 | 571 | 3673003 | ||
BC053085.1 | - | 3 | 570 | 5708827 | ||
U11493.1 | - | 1 | 30 | |||
Z49086.1 | - | 3 | 428 |