Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252188.1 | NP_001239117.1 | 3 | 818 | ||
NM_001252189.1 | NP_001239118.1 | 2 | 745 | |||
NM_010148.3 | NP_034278.1 | 2 | 745 | |||
GenBank mRNA | AF057286.1 | - | 1 | 236 | ||
AK036331.1 | - | 2 | 719 | |||
AK139234.1 | - | 3 | 806 | |||
AK139793.1 | - | 2 | 704 | |||
AK160782.1 | - | 2 | 746 | |||
AK173093.1 | - | 3 | 794 | |||
AK194959.1 | - | 1 | 43 | |||
BC039138.1 | - | 2 | 709 | 2647379 |