Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011879.2 | NP_036009.1 | 5 | 441 | ||
GenBank mRNA | AJ006130.1 | - | 5 | 324 | ||
AK029644.1 | - | 5 | 395 | |||
AK050625.1 | - | 5 | 399 | |||
AK082356.1 | - | 5 | 373 | |||
AK087976.1 | - | 5 | 387 | |||
AK088908.1 | - | 5 | 376 | |||
AK163766.1 | - | 5 | 395 | |||
AK167238.1 | - | 5 | 406 | |||
BC006706.1 | - | 5 | 419 | |||
BC014739.1 | - | 5 | 372 | 4920184 | ||
BC026569.1 | - | 5 | 372 | 4982612 | ||
BC029148.1 | - | 5 | 414 | |||
BC029671.1 | - | 5 | 384 | 5364953 | ||
BC029694.1 | - | 5 | 381 | 5369477 | ||
BC033356.1 | - | 5 | 368 | 5366230 | ||
BG084120.2 | - | 5 | 441 |