Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011879.2 | NP_036009.1 | 16 | 1514 | ||
GenBank mRNA | AJ006130.1 | - | 16 | 1397 | ||
AK082356.1 | - | 16 | 1446 | |||
AK087976.1 | - | 16 | 1460 | |||
BC006706.1 | - | 14 | 1872 | |||
BC014739.1 | - | 16 | 1445 | 4920184 | ||
BC026569.1 | - | 16 | 1445 | 4982612 | ||
BC029148.1 | - | 14 | 2408 | |||
BC029671.1 | - | 16 | 1457 | 5364953 | ||
BC029694.1 | - | 16 | 1454 | 5369477 | ||
BC033356.1 | - | 16 | 1441 | 5366230 |